The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 9 - 10 | 1468 | 63 |
NM_001042495.1 | NP_001035960.1 | 9 - 10 | 1143 | 63 | |
NM_001042496.1 | NP_001035961.1 | 9 - 10 | 1534 | 63 | |
NM_001042497.1 | NP_001035962.1 | 7 - 8 | 1237 | 63 | |
NM_005135.2 | NP_005126.1 | 8 - 9 | 1107 | 63 | |
NM_133647.1 | NP_598408.1 | 8 - 9 | 1282 | 63 | |
XM_006720793.3 | XP_006720856.1 | 7 - 8 | 2017 | 63 | |
XM_011522269.2 | XP_011520571.1 | 8 - 9 | 2164 | 63 | |
GenBank mRNA | AF108831.1 | - | 8 - 9 | 1016 | 63 |
AF116242.1 | - | 8 - 9 | 1118 | 63 | |
AK128133.1 | - | 7 - 8 | 790 | 63 | |
AK292550.1 | - | 9 - 10 | 1221 | 63 | |
AK315283.1 | - | 8 - 9 | 1079 | 63 | |
AL117500.1 | - | 5 - 6 | 710 | 63 | |
BC051744.1 | - | 9 - 10 | 1468 | 63 | |
BC070107.1 | - | 9 - 10 | 1138 | 63 | |
BC126241.1 | - | 8 - 9 | 1015 | 63 | |
BC126243.1 | - | 8 - 9 | 1015 | 63 | |
BX648195.1 | - | 8 - 9 | 2224 | 63 |