Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
Chromosome Location
Chr.19: 16197854 - 16235345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130524.1 NP_001123996.1 12 - 13 1460 66
NM_032493.3 NP_115882.1 11 - 12 1424 66
XM_017027420.1 XP_016882909.1 12 - 13 1400 66
XM_017027421.1 XP_016882910.1 11 - 12 1364 66
GenBank mRNA AB209808.1 - 12 - 13 1380 66
AF290613.1 - 11 - 12 1327 66
AK027528.1 - 11 - 12 1332 66
AK293188.1 - 10 - 11 1208 66
AK297824.1 - 10 - 11 1208 66
BC017469.2 - 11 - 12 1369 66

View Full Product Details