The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130065.1 | NP_001123537.1 | 13 - 14 | 2293 | 120 |
NM_004145.3 | NP_004136.2 | 13 - 14 | 2293 | 120 | |
GenBank mRNA | AB209997.1 | - | 13 - 14 | 2288 | 120 |
AB290184.1 | - | 13 - 14 | 2288 | 120 |