The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130065.1 | NP_001123537.1 | 35 - 36 | 5778 | 100 |
NM_004145.3 | NP_004136.2 | 35 - 36 | 5778 | 100 | |
GenBank mRNA | AF143684.1 | - | 3 - 4 | 340 | 100 |
AK002201.1 | - | 1 - 2 | 145 | 100 | |
BC018108.1 | - | 6 - 7 | 660 | 100 | |
DR006748.1 | - | 4 - 5 | 415 | 100 |