Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
Chromosome Location
Chr.17: 37375710 - 37392708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321399.1 NP_001308328.1 1 - 2 158 76
NM_173625.4 NP_775896.3 1 - 2 158 76
XM_011524648.2 XP_011522950.1 1 - 2 432 76
XM_011524649.2 XP_011522951.1 1 - 2 432 76
GenBank mRNA AK096966.1 - 1 - 2 159 76
BC034672.1 - 1 - 2 101 76

View Full Product Details