The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003742.2 | NP_003733.2 | 13 - 14 | 1562 | 74 |
XM_006712817.3 | XP_006712880.1 | 13 - 14 | 1674 | 74 | |
XM_011512077.2 | XP_011510379.1 | 13 - 14 | 1734 | 74 | |
XM_011512078.2 | XP_011510380.1 | 13 - 14 | 1734 | 74 | |
XM_011512080.2 | XP_011510382.1 | 13 - 14 | 1737 | 74 | |
XM_017005165.1 | XP_016860654.1 | 13 - 14 | 1736 | 74 | |
XM_017005166.1 | XP_016860655.1 | 7 - 8 | 860 | 74 | |
XM_017005167.1 | XP_016860656.1 | 3 - 4 | 418 | 74 | |
GenBank mRNA | AF091582.1 | - | 13 - 14 | 1562 | 74 |
AF136523.1 | - | 13 - 14 | 1563 | 74 |