Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIAA0430
Entrez Gene ID
9665
Gene Name
KIAA0430
Gene Aliases
LKAP, MARF1, PPP1R34
Chromosome Location
Chr.16: 15594369 - 15643166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184998.1 NP_001171927.1 14 - 15 3163 104
NM_001184999.1 NP_001171928.1 14 - 15 3154 104
NM_014647.3 NP_055462.2 14 - 15 3163 104
XM_005255704.1 XP_005255761.1 14 - 15 3159 104
XM_005255706.1 XP_005255763.1 14 - 15 3153 104
XM_005255707.1 XP_005255764.1 14 - 15 3129 104
XM_017023900.1 XP_016879389.1 14 - 15 3047 104
XM_017023901.1 XP_016879390.1 14 - 15 3044 104
XM_017023902.1 XP_016879391.1 13 - 14 2997 104
XM_017023903.1 XP_016879392.1 13 - 14 2730 104
XM_017023904.1 XP_016879393.1 13 - 14 2727 104
XM_017023905.1 XP_016879394.1 14 - 15 2584 104
XM_017023906.1 XP_016879395.1 14 - 15 2525 104
GenBank mRNA AB007890.3 - 12 - 13 2249 104
BC137165.1 - 14 - 15 3105 104
BC137170.1 - 14 - 15 3105 104
BC144514.1 - 14 - 15 3105 104
BC144515.1 - 14 - 15 3096 104
CN310535.1 - 1 - 2 242 104

View Full Product Details