Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA0430
Entrez Gene ID
9665
Gene Name
KIAA0430
Gene Aliases
LKAP, MARF1, PPP1R34
Chromosome Location
Chr.16: 15594369 - 15643166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184998.1 NP_001171927.1 17 - 18 3679 82
NM_001184999.1 NP_001171928.1 17 - 18 3670 82
NM_014647.3 NP_055462.2 17 - 18 3679 82
XM_005255704.1 XP_005255761.1 17 - 18 3675 82
XM_005255706.1 XP_005255763.1 17 - 18 3669 82
XM_005255707.1 XP_005255764.1 17 - 18 3645 82
XM_017023900.1 XP_016879389.1 17 - 18 3563 82
XM_017023901.1 XP_016879390.1 17 - 18 3560 82
XM_017023902.1 XP_016879391.1 16 - 17 3513 82
XM_017023903.1 XP_016879392.1 16 - 17 3246 82
XM_017023904.1 XP_016879393.1 16 - 17 3243 82
XM_017023905.1 XP_016879394.1 17 - 18 3100 82
XM_017023906.1 XP_016879395.1 17 - 18 3041 82
GenBank mRNA AB007890.3 - 15 - 16 2765 82
BC137165.1 - 17 - 18 3621 82
BC137170.1 - 17 - 18 3621 82
BC144514.1 - 17 - 18 3621 82
BC144515.1 - 17 - 18 3612 82

View Full Product Details