The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137550.1 | NP_001131022.1 | 11 - 12 | 697 | 118 |
XM_017005253.1 | XP_016860742.1 | 12 - 13 | 759 | 118 | |
XM_017005254.1 | XP_016860743.1 | 12 - 13 | 759 | 118 | |
XM_017005255.1 | XP_016860744.1 | 12 - 13 | 759 | 118 | |
XM_017005256.1 | XP_016860745.1 | 12 - 13 | 759 | 118 | |
XM_017005257.1 | XP_016860746.1 | 11 - 12 | 696 | 118 | |
XM_017005258.1 | XP_016860747.1 | 12 - 13 | 759 | 118 | |
XM_017005259.1 | XP_016860748.1 | 8 - 9 | 558 | 118 | |
XM_017005260.1 | XP_016860749.1 | 10 - 11 | 657 | 118 | |
GenBank mRNA | AF115510.1 | - | 11 - 12 | 566 | 118 |
BG699031.1 | - | 8 - 9 | 558 | 119 |