Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRFIP1
Entrez Gene ID
9208
Gene Name
LRR binding FLII interacting protein 1
Gene Aliases
FLAP-1, FLAP1, FLIIAP1, GCF-2, GCF2, HUFI-1, TRIP
Chromosome Location
Chr.2: 237627570 - 237781647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137550.1 NP_001131022.1 4 - 5 308 121
XM_017005253.1 XP_016860742.1 4 - 5 307 121
XM_017005254.1 XP_016860743.1 4 - 5 307 121
XM_017005255.1 XP_016860744.1 4 - 5 307 121
XM_017005256.1 XP_016860745.1 4 - 5 307 121
XM_017005257.1 XP_016860746.1 4 - 5 307 121
XM_017005258.1 XP_016860747.1 4 - 5 307 121
XM_017005259.1 XP_016860748.1 4 - 5 319 121
XM_017005260.1 XP_016860749.1 4 - 5 307 121
XM_017005261.1 XP_016860750.1 4 - 5 307 121
XM_017005262.1 XP_016860751.1 4 - 5 307 121
GenBank mRNA AF115510.1 - 4 - 5 177 121
BG699031.1 - 4 - 5 318 121

View Full Product Details