The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000111.2 | NP_000102.1 | 17 - 18 | 2220 | 127 |
XM_011515867.2 | XP_011514169.1 | 17 - 18 | 2201 | 127 | |
GenBank mRNA | AK300651.1 | - | 11 - 12 | 1248 | 127 |
BC025671.1 | - | 17 - 18 | 2194 | 127 | |
L02785.1 | - | 17 - 18 | 2193 | 127 |