The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198810.1 | NP_001185739.1 | 1 - 2 | 106 | 83 |
XM_011545321.2 | XP_011543623.1 | 1 - 2 | 120 | 83 | |
GenBank mRNA | AF045584.1 | - | 1 - 2 | 64 | 83 |
BC001639.2 | - | 1 - 2 | 106 | 83 |