Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAAF1
Entrez Gene ID
80227
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
PAAF, Rpn14, WDR71
Chromosome Location
Chr.11: 73876699 - 73927736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267803.1 NP_001254732.1 6 - 7 497 133
NM_001267804.1 NP_001254733.1 6 - 7 488 133
NM_001267805.1 NP_001254734.1 6 - 7 484 133
NM_001267806.1 NP_001254735.1 5 - 6 371 133
NM_025155.2 NP_079431.1 5 - 6 428 133
XM_005274308.2 XP_005274365.2 3 - 4 1020 133
GenBank mRNA AF087895.1 - 6 - 7 422 133
AK021910.1 - 5 - 6 409 133
AK222501.1 - 5 - 6 409 133
AK225520.1 - 5 - 6 396 133
AK298258.1 - 5 - 6 371 133
AK309133.1 - 3 - 4 635 133
AK316394.1 - 5 - 6 642 133
BC006142.2 - 5 - 6 381 133
BC021541.1 - 5 - 6 392 133
BC028628.1 - 6 - 7 462 133
CD243380.1 - 6 - 7 474 133
DC354686.1 - 5 - 6 430 133

View Full Product Details