Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAAF1
Entrez Gene ID
80227
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
PAAF, Rpn14, WDR71
Chromosome Location
Chr.11: 73876699 - 73927736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267803.1 NP_001254732.1 9 - 10 941 67
NM_001267804.1 NP_001254733.1 9 - 10 932 67
NM_001267805.1 NP_001254734.1 9 - 10 928 67
NM_001267806.1 NP_001254735.1 8 - 9 815 67
NM_025155.2 NP_079431.1 8 - 9 872 67
XM_005274308.2 XP_005274365.2 6 - 7 1464 67
XM_005274309.1 XP_005274366.1 7 - 8 731 67
XM_017018357.1 XP_016873846.1 9 - 10 860 67
XM_017018358.1 XP_016873847.1 9 - 10 865 67
XM_017018359.1 XP_016873848.1 6 - 7 1423 67
GenBank mRNA AF087895.1 - 9 - 10 866 67
AK021910.1 - 8 - 9 853 67
AK225520.1 - 8 - 9 840 67
AK298258.1 - 8 - 9 815 67
AK309133.1 - 6 - 7 1079 67
AK316394.1 - 8 - 9 1086 67
BC006142.2 - 8 - 9 825 67
BC021541.1 - 8 - 9 836 67
BC028628.1 - 9 - 10 906 67

View Full Product Details