The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005180.8 | NP_005171.4 | 9 - 10 | 1162 | 153 |
GenBank mRNA | BC011652.2 | - | 9 - 10 | 1161 | 153 |
BT019450.1 | - | 8 - 9 | 656 | 153 | |
CR456911.1 | - | 8 - 9 | 656 | 153 | |
L13689.1 | - | 9 - 10 | 1135 | 153 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204062.1 | NP_001190991.1 | 13 - 14 | 1120 | 153 |
GenBank mRNA | AK313235.1 | - | 13 - 14 | 1095 | 153 |