Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204197.1 NP_001191126.1 2 - 3 201 132
NM_015678.4 NP_056493.3 42 - 43 7231 132
XM_005266346.4 XP_005266403.1 42 - 43 7231 132
XM_005266347.4 XP_005266404.2 42 - 43 7231 132
XM_005266348.4 XP_005266405.2 41 - 42 7222 132
XM_006719803.3 XP_006719866.1 42 - 43 7231 132
XM_006719805.3 XP_006719868.1 41 - 42 7222 132
XM_006719806.3 XP_006719869.1 41 - 42 7222 132
XM_011535046.2 XP_011533348.1 34 - 35 5362 132
XM_011535047.2 XP_011533349.1 42 - 43 7231 132
XM_017020544.1 XP_016876033.1 42 - 43 7213 132
XM_017020545.1 XP_016876034.1 42 - 43 7213 132
XM_017020546.1 XP_016876035.1 42 - 43 6519 132
GenBank mRNA AB046764.1 - 9 - 10 943 132
AF467288.1 - 43 - 44 6903 132
AK091469.1 - 13 - 14 1390 132
AK122888.1 - 12 - 13 1744 132
AK127791.1 - 19 - 20 2582 132
AK294737.1 - 2 - 3 201 132
AK297773.1 - 3 - 4 300 132
BX640760.1 - 18 - 19 2561 132

View Full Product Details