The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193478.1 | NP_001180407.1 | 3 - 4 | 376 | 86 |
NM_030911.3 | NP_112173.1 | 3 - 4 | 376 | 86 | |
XM_011535250.2 | XP_011533552.1 | 3 - 4 | 340 | 86 | |
XM_011535251.1 | XP_011533553.1 | 3 - 4 | 320 | 86 | |
XM_017020779.1 | XP_016876268.1 | 3 - 4 | 368 | 86 | |
XM_017020780.1 | XP_016876269.1 | 3 - 4 | 718 | 86 | |
XM_017020782.1 | XP_016876271.1 | 2 - 3 | 166 | 86 | |
GenBank mRNA | AK312835.1 | - | 3 - 4 | 346 | 86 |
AY027525.1 | - | 3 - 4 | 325 | 86 | |
BC032889.1 | - | 3 - 4 | 351 | 86 | |
BC070238.1 | - | 3 - 4 | 344 | 86 | |
BC092434.1 | - | 3 - 4 | 354 | 86 | |
BC125202.1 | - | 3 - 4 | 290 | 86 | |
DA400049.1 | - | 3 - 4 | 376 | 86 | |
DA560379.1 | - | 3 - 4 | 376 | 86 |