Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 23 - 24 3336 82
XM_006717308.2 XP_006717371.1 22 - 23 3350 82
XM_006717310.3 XP_006717373.1 21 - 22 1149 82
XM_011519138.2 XP_011517440.1 23 - 24 2968 82
XM_011519140.1 XP_011517442.1 23 - 24 3404 82
XM_011519142.2 XP_011517444.1 23 - 24 3410 82
XM_011519143.2 XP_011517445.1 23 - 24 3408 82
XM_011519144.2 XP_011517446.1 23 - 24 3408 82
XM_011519145.2 XP_011517447.1 10 - 11 578 82
XM_017015239.1 XP_016870728.1 23 - 24 3409 82
GenBank mRNA AB058773.1 - 20 - 21 1816 82
AY149237.1 - 23 - 24 2930 82

View Full Product Details