Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 30 - 31 3707 74
XM_006717308.2 XP_006717371.1 29 - 30 3721 74
XM_006717310.3 XP_006717373.1 28 - 29 1520 74
XM_011519138.2 XP_011517440.1 30 - 31 3339 74
XM_011519140.1 XP_011517442.1 29 - 30 3721 74
XM_011519142.2 XP_011517444.1 30 - 31 3781 74
XM_011519143.2 XP_011517445.1 30 - 31 3779 74
XM_011519144.2 XP_011517446.1 30 - 31 3779 74
XM_011519145.2 XP_011517447.1 17 - 18 949 74
XM_017015239.1 XP_016870728.1 30 - 31 3780 74
GenBank mRNA AB058773.1 - 27 - 28 2194 74
AY149237.1 - 30 - 31 3301 74

View Full Product Details