The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032888.3 | NP_116277.2 | 7 - 8 | 2534 | 90 |
XM_006717308.2 | XP_006717371.1 | 6 - 7 | 2548 | 90 | |
XM_006717310.3 | XP_006717373.1 | 5 - 6 | 359 | 90 | |
XM_011519138.2 | XP_011517440.1 | 7 - 8 | 2166 | 90 | |
XM_011519140.1 | XP_011517442.1 | 7 - 8 | 2602 | 90 | |
XM_011519142.2 | XP_011517444.1 | 7 - 8 | 2608 | 90 | |
XM_011519143.2 | XP_011517445.1 | 7 - 8 | 2606 | 90 | |
XM_011519144.2 | XP_011517446.1 | 7 - 8 | 2606 | 90 | |
XM_017015239.1 | XP_016870728.1 | 7 - 8 | 2607 | 90 | |
GenBank mRNA | AB058773.1 | - | 4 - 5 | 1022 | 90 |
AY149237.1 | - | 7 - 8 | 2128 | 90 |