The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002740.5 | NP_002731.4 | 7 - 8 | 879 | 102 |
XM_017006860.1 | XP_016862349.1 | 6 - 7 | 786 | 102 | |
GenBank mRNA | AB451443.1 | - | 7 - 8 | 614 | 102 |
AK315342.1 | - | 7 - 8 | 807 | 102 | |
BC022016.2 | - | 7 - 8 | 874 | 102 | |
L18964.1 | - | 7 - 8 | 878 | 102 | |
L33881.1 | - | 7 - 8 | 818 | 102 |