The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 11 - 12 | 1327 | 80 |
NM_001001486.1 | NP_001001486.1 | 10 - 11 | 1004 | 80 | |
NM_001001487.1 | NP_001001487.1 | 10 - 11 | 1004 | 80 | |
NM_001199179.1 | NP_001186108.1 | 11 - 12 | 1233 | 80 | |
NM_001199180.1 | NP_001186109.1 | 10 - 11 | 1222 | 80 | |
NM_001199181.1 | NP_001186110.1 | 10 - 11 | 1222 | 80 | |
NM_001199182.1 | NP_001186111.1 | 9 - 10 | 1105 | 80 | |
NM_001199183.1 | NP_001186112.1 | 10 - 11 | 1095 | 80 | |
NM_001199184.1 | NP_001186113.1 | 10 - 11 | 1095 | 80 | |
NM_001199185.1 | NP_001186114.1 | 10 - 11 | 1146 | 80 | |
NM_014382.3 | NP_055197.2 | 10 - 11 | 1146 | 80 | |
XM_005247354.2 | XP_005247411.1 | 10 - 11 | 1247 | 80 | |
XM_005247355.2 | XP_005247412.1 | 11 - 12 | 1374 | 80 | |
XM_005247356.2 | XP_005247413.1 | 11 - 12 | 1127 | 80 | |
XM_005247358.2 | XP_005247415.1 | 10 - 11 | 951 | 80 | |
XM_017006164.1 | XP_016861653.1 | 11 - 12 | 1035 | 80 | |
GenBank mRNA | AB037768.1 | - | 11 - 12 | 1234 | 80 |
AF181120.1 | - | 10 - 11 | 1059 | 80 | |
AF181121.1 | - | 10 - 11 | 1059 | 80 | |
AF189723.2 | - | 10 - 11 | 978 | 80 | |
AF225981.1 | - | 10 - 11 | 978 | 80 | |
AK074692.1 | - | 8 - 9 | 685 | 80 | |
AK296470.1 | - | 10 - 11 | 1149 | 80 | |
AK299945.1 | - | 9 - 10 | 861 | 80 | |
AK304175.1 | - | 10 - 11 | 951 | 80 | |
AK304374.1 | - | 10 - 11 | 978 | 80 | |
AK314342.1 | - | 10 - 11 | 1008 | 80 | |
AY268374.1 | - | 10 - 11 | 953 | 80 | |
AY268375.1 | - | 10 - 11 | 953 | 80 | |
BC028139.1 | - | 10 - 11 | 1016 | 80 |