The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003794.2 | NP_001003794.1 | 3 - 4 | 420 | 61 |
NM_001256585.1 | NP_001243514.1 | 3 - 4 | 827 | 61 | |
NM_007283.6 | NP_009214.1 | 3 - 4 | 827 | 61 | |
XM_011512377.1 | XP_011510679.1 | 3 - 4 | 799 | 61 | |
XM_011512378.1 | XP_011510680.1 | 3 - 4 | 420 | 61 | |
XM_011512382.1 | XP_011510684.1 | 2 - 3 | 362 | 61 | |
XM_017005662.1 | XP_016861151.1 | 3 - 4 | 420 | 61 | |
XM_017005666.1 | XP_016861155.1 | 3 - 4 | 420 | 61 | |
GenBank mRNA | AJ270950.1 | - | 2 - 3 | 233 | 61 |
AK091314.1 | - | 3 - 4 | 1130 | 61 | |
AK304844.1 | - | 3 - 4 | 296 | 61 | |
AK315529.1 | - | 3 - 4 | 467 | 61 | |
BC000551.2 | - | 3 - 4 | 412 | 61 | |
BC006230.2 | - | 3 - 4 | 782 | 61 | |
BE302457.1 | - | 3 - 4 | 369 | 61 | |
BG249567.1 | - | - | 378 | 61 | |
BP307995.1 | - | 3 - 4 | 398 | 61 | |
CR456835.1 | - | 2 - 3 | 263 | 61 | |
DA240965.1 | - | 3 - 4 | 420 | 61 | |
EU660904.1 | - | 2 - 3 | 233 | 61 | |
U67963.1 | - | 3 - 4 | 413 | 61 |