The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033429.1 | 1 - 2 | 143 | 86 | |
XM_017007286.1 | XP_016862775.1 | 1 - 2 | 140 | 86 | |
GenBank mRNA | AB053326.1 | - | 1 - 2 | 122 | 86 |