Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABTB1
Entrez Gene ID
80325
Gene Name
ankyrin repeat and BTB domain containing 1
Gene Aliases
BPOZ, BTB3, BTBD21, EF1ABP, PP2259
Chromosome Location
Chr.3: 127672928 - 127680926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033429.1 2 - 3 172 65
XM_017007286.1 XP_016862775.1 2 - 3 169 65
GenBank mRNA AB053326.1 - 2 - 3 151 65

View Full Product Details