The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007027.3 | NP_008958.2 | 2 - 3 | 219 | 152 |
XM_005247076.3 | XP_005247133.1 | 2 - 3 | 706 | 152 | |
XM_011512357.2 | XP_011510659.1 | 2 - 3 | 290 | 152 | |
XM_017005636.1 | XP_016861125.1 | 2 - 3 | 289 | 152 | |
GenBank mRNA | AB019397.1 | - | 2 - 3 | 166 | 152 |
AK302584.1 | - | 2 - 3 | 185 | 152 | |
BC032633.1 | - | 2 - 3 | 230 | 152 | |
BC126209.1 | - | 1 - 2 | 109 | 152 | |
BC151237.1 | - | 2 - 3 | 172 | 152 | |
D87448.1 | - | 2 - 3 | 172 | 152 |