The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007027.3 | NP_008958.2 | 4 - 5 | 495 | 118 |
XM_005247076.3 | XP_005247133.1 | 4 - 5 | 982 | 118 | |
XM_011512357.2 | XP_011510659.1 | 4 - 5 | 566 | 118 | |
XM_017005636.1 | XP_016861125.1 | 4 - 5 | 565 | 118 | |
XM_017005637.1 | XP_016861126.1 | 1 - 2 | 101 | 118 | |
GenBank mRNA | AB019397.1 | - | 4 - 5 | 442 | 118 |
AK302584.1 | - | 4 - 5 | 461 | 118 | |
BC032633.1 | - | 4 - 5 | 506 | 118 | |
BC126209.1 | - | 3 - 4 | 385 | 118 | |
BC151237.1 | - | 4 - 5 | 448 | 118 | |
D87448.1 | - | 4 - 5 | 448 | 118 |