The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007027.3 | NP_008958.2 | 6 - 7 | 870 | 111 |
XM_005247076.3 | XP_005247133.1 | 6 - 7 | 1357 | 111 | |
XM_011512357.2 | XP_011510659.1 | 6 - 7 | 941 | 111 | |
XM_017005636.1 | XP_016861125.1 | 6 - 7 | 940 | 111 | |
XM_017005637.1 | XP_016861126.1 | 3 - 4 | 476 | 111 | |
GenBank mRNA | AB019397.1 | - | 6 - 7 | 817 | 111 |
AK302584.1 | - | 6 - 7 | 836 | 111 | |
BC126209.1 | - | 5 - 6 | 760 | 111 | |
BC151237.1 | - | 6 - 7 | 823 | 111 | |
D87448.1 | - | 6 - 7 | 823 | 111 |