The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199461.3 | NP_955631.1 | 1 - 1 | 1953 | 85 |
GenBank mRNA | AF458985.1 | - | 1 - 1 | 1953 | 85 |