Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
Chromosome Location
Chr.1: 186375758 - 186421375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164245.1 NP_001157717.1 9 - 10 1076 79
NM_001164246.1 NP_001157718.1 9 - 10 1049 79
NM_017847.5 NP_060317.3 10 - 11 1145 79
XM_011509660.1 XP_011507962.1 10 - 11 1144 79
XM_011509661.1 XP_011507963.1 10 - 11 1144 79
GenBank mRNA AK000512.1 - 10 - 11 1065 79
AK026086.1 - 8 - 9 715 79
AK298107.1 - 9 - 10 985 79
AY854248.1 - 10 - 11 1038 79
BC003397.1 - 10 - 11 978 79
DQ323046.1 - 9 - 10 909 79
DQ529299.1 - 8 - 9 840 79

View Full Product Details