The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164245.1 | NP_001157717.1 | 6 - 7 | 708 | 107 |
NM_001164246.1 | NP_001157718.1 | 5 - 6 | 612 | 107 | |
NM_017847.5 | NP_060317.3 | 6 - 7 | 708 | 107 | |
XM_011509660.1 | XP_011507962.1 | 6 - 7 | 707 | 107 | |
XM_011509661.1 | XP_011507963.1 | 6 - 7 | 707 | 107 | |
XM_017001543.1 | XP_016857032.1 | 6 - 7 | 707 | 107 | |
GenBank mRNA | AK000512.1 | - | 6 - 7 | 628 | 107 |
AK026086.1 | - | 4 - 5 | 278 | 107 | |
AK298107.1 | - | 5 - 6 | 548 | 107 | |
AY854248.1 | - | 6 - 7 | 601 | 107 | |
BC003397.1 | - | 6 - 7 | 541 | 107 | |
DQ323046.1 | - | 5 - 6 | 472 | 107 |