The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164246.1 | NP_001157718.1 | 7 - 8 | 841 | 149 |
NM_017847.5 | NP_060317.3 | 8 - 9 | 937 | 149 | |
XM_011509660.1 | XP_011507962.1 | 8 - 9 | 936 | 149 | |
XM_011509661.1 | XP_011507963.1 | 8 - 9 | 936 | 149 | |
XM_017001543.1 | XP_016857032.1 | 8 - 9 | 936 | 149 | |
GenBank mRNA | AK000512.1 | - | 8 - 9 | 857 | 149 |
AK026086.1 | - | 6 - 7 | 507 | 149 | |
AK298107.1 | - | 7 - 8 | 777 | 149 | |
AY854248.1 | - | 8 - 9 | 830 | 149 | |
BC003397.1 | - | 8 - 9 | 770 | 149 | |
DQ323046.1 | - | 7 - 8 | 701 | 149 |