The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001263.3 | NP_001254.2 | 1 - 2 | 615 | 109 |
XM_005262687.2 | XP_005262744.1 | 1 - 2 | 611 | 109 | |
XM_017007648.1 | XP_016863137.1 | 1 - 2 | 611 | 109 | |
XM_017007649.1 | XP_016863138.1 | 1 - 2 | 611 | 109 | |
XM_017007650.1 | XP_016863139.1 | 1 - 2 | 611 | 109 | |
XM_017007651.1 | XP_016863140.1 | 1 - 2 | 611 | 109 | |
GenBank mRNA | AK314245.1 | - | 1 - 2 | 354 | 109 |
BC074833.2 | - | 1 - 2 | 166 | 109 | |
BC074881.2 | - | 1 - 2 | 166 | 109 | |
U60808.1 | - | 1 - 2 | 266 | 109 | |
U65887.1 | - | 1 - 2 | 553 | 109 |