Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCHY1
Entrez Gene ID
25898
Gene Name
ring finger and CHY zinc finger domain containing 1
Gene Aliases
ARNIP, CHIMP, PIRH2, PRO1996, RNF199, ZCHY, ZNF363
Chromosome Location
Chr.4: 75479037 - 75515057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009922.2 NP_001009922.1 5 - 6 594 69
NM_001278536.1 NP_001265465.1 4 - 5 474 69
NM_001278537.1 NP_001265466.1 4 - 5 474 69
NM_001278538.1 NP_001265467.1 5 - 6 550 69
NM_001278539.1 NP_001265468.1 5 - 6 632 69
NM_015436.3 NP_056251.2 5 - 6 594 69
NR_037913.1 5 - 6 594 69
NR_037914.1 5 - 6 590 69
NR_103723.1 5 - 6 594 69
NR_103724.1 5 - 6 594 69
NR_103725.1 4 - 5 368 69
XM_011531838.2 XP_011530140.1 5 - 6 563 69
XM_011531839.2 XP_011530141.1 5 - 6 751 69
GenBank mRNA AB209072.1 - 5 - 6 457 69
AF247041.1 - 5 - 6 450 69
AF255666.1 - 5 - 6 476 69
AF305424.1 - 5 - 6 484 69
AK091501.1 - 5 - 6 565 69
AK308754.1 - 4 - 5 327 69
AK308848.1 - 2 - 3 376 69
AK309555.1 - 5 - 6 437 69
AL050144.1 - 4 - 5 351 69
AY888047.1 - 5 - 6 476 69
BC015464.1 - 1 - 2 46 69
BG703058.1 - 5 - 6 463 69
BP314571.1 - 4 - 5 341 69
BQ959587.1 - 5 - 6 423 69
CF597260.1 - 3 - 4 165 69
GQ250944.1 - 5 - 6 450 69
GQ250945.1 - 5 - 6 450 69
GQ250946.1 - 5 - 6 450 69
GU937000.1 - 5 - 6 478 69
JF691538.1 - 5 - 6 450 69
JF691539.1 - 5 - 6 450 69

View Full Product Details