Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCHY1
Entrez Gene ID
25898
Gene Name
ring finger and CHY zinc finger domain containing 1
Gene Aliases
ARNIP, CHIMP, PIRH2, PRO1996, RNF199, ZCHY, ZNF363
Chromosome Location
Chr.4: 75479037 - 75515057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278536.1 NP_001265465.1 5 - 6 530 87
NM_001278538.1 NP_001265467.1 6 - 7 606 87
NM_001278539.1 NP_001265468.1 6 - 7 688 87
NM_015436.3 NP_056251.2 6 - 7 650 87
XM_011531838.2 XP_011530140.1 6 - 7 619 87
XM_011531839.2 XP_011530141.1 6 - 7 807 87
GenBank mRNA AB209072.1 - 6 - 7 513 87
AF247041.1 - 6 - 7 506 87
AF255666.1 - 6 - 7 532 87
AF305424.1 - 6 - 7 540 87
AK091501.1 - 6 - 7 621 87
AK308754.1 - 5 - 6 383 87
AK308848.1 - 3 - 4 432 87
AL050144.1 - 5 - 6 407 87
BC047393.1 - 6 - 7 650 87
BG703058.1 - 6 - 7 519 87
BQ959587.1 - 6 - 7 479 87
GQ250944.1 - 6 - 7 506 87
GU937000.1 - 6 - 7 534 87
JF691538.1 - 6 - 7 506 87
JF691539.1 - 6 - 7 506 87

View Full Product Details