Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADH5
Entrez Gene ID
128
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Gene Aliases
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p
Chromosome Location
Chr.4: 99070978 - 99088788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000671.4 NP_000662.3 6 - 7 914 82
GenBank mRNA AK312367.1 - 6 - 7 856 82
BC014665.1 - 5 - 6 828 82
BC070491.1 - 6 - 7 848 82
BT019832.1 - 5 - 6 826 82
BT019833.1 - 5 - 6 826 82
CR536491.1 - 5 - 6 826 82
CR541689.1 - 5 - 6 826 82
GQ472236.1 - 5 - 6 828 82
M29872.1 - 6 - 7 884 82
M30471.1 - 5 - 6 828 82

View Full Product Details