The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000671.4 | NP_000662.3 | 7 - 8 | 1049 | 166 |
GenBank mRNA | AK312367.1 | - | 7 - 8 | 991 | 166 |
BC014665.1 | - | 6 - 7 | 963 | 166 | |
BC070491.1 | - | 7 - 8 | 983 | 166 | |
BT019832.1 | - | 6 - 7 | 961 | 166 | |
BT019833.1 | - | 6 - 7 | 961 | 166 | |
CR536491.1 | - | 6 - 7 | 961 | 166 | |
CR541689.1 | - | 6 - 7 | 961 | 166 | |
GQ472236.1 | - | 6 - 7 | 963 | 166 | |
M29872.1 | - | 7 - 8 | 1019 | 166 | |
M30471.1 | - | 6 - 7 | 963 | 166 |