The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134707.1 | NP_001128179.1 | 13 - 14 | 1801 | 68 |
NM_007101.3 | NP_009032.2 | 13 - 14 | 1960 | 68 | |
XM_017014367.1 | XP_016869856.1 | 13 - 14 | 1963 | 68 | |
XM_017014368.1 | XP_016869857.1 | 13 - 14 | 1963 | 68 | |
GenBank mRNA | AF047190.1 | - | 13 - 14 | 1801 | 68 |
AF095735.2 | - | 13 - 14 | 1925 | 68 | |
AF129265.1 | - | 9 - 10 | 953 | 68 | |
AF162428.1 | - | 12 - 13 | 1697 | 68 | |
AJ223317.1 | - | 13 - 14 | 1801 | 68 | |
AK093794.1 | - | 1 - 2 | 785 | 68 | |
AK223402.1 | - | 13 - 14 | 1847 | 68 | |
AK291211.1 | - | 13 - 14 | 1795 | 68 | |
AK298348.1 | - | 13 - 14 | 1844 | 68 | |
AK316494.1 | - | 12 - 13 | 1599 | 68 | |
BC033217.1 | - | - | 598 | 68 | |
BC041574.1 | - | 6 - 7 | 607 | 68 | |
BC136363.1 | - | 13 - 14 | 1778 | 68 | |
BC136364.1 | - | 13 - 14 | 1778 | 68 | |
BC144035.1 | - | 13 - 14 | 1796 | 68 |