The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080424.1 | NP_001073893.1 | 9 - 10 | 1286 | 82 |
XM_005256549.3 | XP_005256606.1 | 10 - 11 | 1438 | 82 | |
XM_005256550.4 | XP_005256607.1 | 9 - 10 | 1280 | 82 | |
XM_005256551.3 | XP_005256608.1 | 9 - 10 | 1319 | 82 | |
XM_005256552.4 | XP_005256609.1 | 9 - 10 | 2292 | 82 | |
XM_006721483.3 | XP_006721546.1 | 10 - 11 | 1383 | 82 | |
XM_011523750.2 | XP_011522052.1 | 10 - 11 | 2643 | 82 | |
XM_011523752.2 | XP_011522054.1 | 9 - 10 | 1283 | 82 | |
XM_017024381.1 | XP_016879870.1 | 9 - 10 | 2729 | 82 | |
GenBank mRNA | AB002344.2 | - | 9 - 10 | 1291 | 82 |