Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPH1
Entrez Gene ID
65059
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Gene Aliases
ALS2CR18, ALS2CR9, LPD, PREL-2, PREL2, RMO1, RalGDS/AF-6
Chromosome Location
Chr.2: 203433682 - 203535335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203365.3 NP_976241.1 8 - 9 1352 109
NM_213589.2 NP_998754.1 6 - 7 1196 109
XM_005246777.3 XP_005246834.1 8 - 9 1347 109
XM_005246779.4 XP_005246836.1 8 - 9 2246 109
XM_006712695.3 XP_006712758.1 9 - 10 2327 109
XM_011511646.2 XP_011509948.1 9 - 10 1419 109
XM_011511647.1 XP_011509949.1 6 - 7 1052 109
XM_011511648.1 XP_011509950.1 6 - 7 1046 109
XM_011511649.2 XP_011509951.1 8 - 9 1347 109
GenBank mRNA AB051468.1 - 5 - 6 929 109
AB053311.1 - 8 - 9 1346 109
AJ584699.1 - 6 - 7 1190 109
AY494951.1 - 6 - 7 1192 109
AY523977.1 - 8 - 9 1348 109
AY523978.1 - 8 - 9 1348 109

View Full Product Details