The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206846.1 | NP_001193775.1 | 7 - 8 | 2740 | 106 |
NM_018136.4 | NP_060606.3 | 7 - 8 | 2740 | 106 | |
GenBank mRNA | AF509326.1 | - | 7 - 8 | 2483 | 106 |
AK001380.1 | - | 3 - 4 | 415 | 106 | |
AK125107.1 | - | 7 - 8 | 2740 | 106 | |
AK226178.1 | - | 7 - 8 | 2673 | 106 | |
AY099891.1 | - | 5 - 6 | 1265 | 106 | |
AY099893.1 | - | 5 - 6 | 1265 | 106 | |
AY367065.1 | - | 7 - 8 | 2483 | 106 | |
AY971956.1 | - | 7 - 8 | 2483 | 106 | |
BC034607.1 | - | 3 - 4 | 359 | 106 |