Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPM
Entrez Gene ID
259266
Gene Name
abnormal spindle microtubule assembly
Gene Aliases
ASP, Calmbp1, MCPH5
Chromosome Location
Chr.1: 197084127 - 197146694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206846.1 NP_001193775.1 19 - 20 4588 158
NM_018136.4 NP_060606.3 20 - 21 9343 158
GenBank mRNA AF509326.1 - 20 - 21 9086 158
AK001379.1 - 3 - 4 2477 158
AK001411.1 - 3 - 4 870 158
AK226178.1 - 19 - 20 4521 157
AY367065.1 - 20 - 21 9086 158
AY971955.1 - 6 - 7 1841 158
AY971956.1 - 19 - 20 4331 158
AY971957.1 - 6 - 7 3044 158
BC034607.1 - 15 - 16 2207 158
BC040439.1 - 3 - 4 905 158
BX648804.1 - 3 - 4 4197 158

View Full Product Details