The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206846.1 | NP_001193775.1 | 26 - 27 | 5829 | 115 |
NM_018136.4 | NP_060606.3 | 27 - 28 | 10584 | 115 | |
GenBank mRNA | AF509326.1 | - | 27 - 28 | 10327 | 115 |
AK001379.1 | - | 10 - 11 | 3718 | 115 | |
AK001411.1 | - | 10 - 11 | 2111 | 115 | |
AK226178.1 | - | 26 - 27 | 5761 | 115 | |
AY367065.1 | - | 27 - 28 | 10327 | 115 | |
AY971955.1 | - | 13 - 14 | 3082 | 115 | |
AY971956.1 | - | 26 - 27 | 5572 | 115 | |
BC034607.1 | - | 22 - 23 | 3448 | 115 | |
BC040439.1 | - | 10 - 11 | 2146 | 115 | |
BX648804.1 | - | 10 - 11 | 5438 | 115 |