The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195215.1 | NP_001182144.1 | 19 - 20 | 1742 | 127 |
NR_125340.1 | 16 - 17 | 1489 | 127 | ||
XM_005244931.2 | XP_005244988.1 | 18 - 19 | 1682 | 127 | |
XM_006711192.2 | XP_006711255.1 | 20 - 21 | 1652 | 127 | |
XM_006711193.3 | XP_006711256.1 | 20 - 21 | 1784 | 127 | |
XM_006711194.3 | XP_006711257.1 | 7 - 8 | 11780 | 127 | |
XM_011509246.2 | XP_011507548.1 | 19 - 20 | 1603 | 127 | |
XM_011509247.1 | XP_011507549.1 | 18 - 19 | 1543 | 127 | |
XM_011509248.2 | XP_011507550.1 | 19 - 20 | 1536 | 127 | |
XM_011509249.2 | XP_011507551.1 | 20 - 21 | 1841 | 127 | |
XM_017000470.1 | XP_016855959.1 | 20 - 21 | 1730 | 127 | |
XM_017000471.1 | XP_016855960.1 | 19 - 20 | 1592 | 127 | |
XM_017000472.1 | XP_016855961.1 | 15 - 16 | 1216 | 127 | |
GenBank mRNA | AK000061.1 | - | 5 - 6 | 279 | 127 |
AK091207.1 | - | 16 - 17 | 1303 | 127 | |
BC074735.2 | - | 2 - 3 | 128 | 127 | |
BP220095.1 | - | 5 - 6 | 279 | 127 |