The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031746.4 | NP_001026916.2 | 3 - 4 | 577 | 130 |
XM_017015825.1 | XP_016871314.1 | 3 - 4 | 617 | 130 | |
XM_017015826.1 | XP_016871315.1 | 3 - 4 | 617 | 130 | |
XM_017015827.1 | XP_016871316.1 | 3 - 4 | 557 | 130 | |
XM_017015828.1 | XP_016871317.1 | 3 - 4 | 997 | 130 | |
GenBank mRNA | BC041414.1 | - | 3 - 4 | 559 | 130 |