The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031746.4 | NP_001026916.2 | 4 - 5 | 677 | 70 |
XM_017015825.1 | XP_016871314.1 | 4 - 5 | 717 | 70 | |
XM_017015826.1 | XP_016871315.1 | 4 - 5 | 717 | 70 | |
XM_017015827.1 | XP_016871316.1 | 4 - 5 | 657 | 70 | |
XM_017015828.1 | XP_016871317.1 | 4 - 5 | 1097 | 70 | |
GenBank mRNA | AK297931.1 | - | 4 - 5 | 652 | 70 |
BC041414.1 | - | 4 - 5 | 659 | 70 |