The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031746.4 | NP_001026916.2 | 7 - 8 | 889 | 134 |
XM_017015828.1 | XP_016871317.1 | 7 - 8 | 1309 | 134 | |
GenBank mRNA | AK297931.1 | - | 7 - 8 | 864 | 134 |
BC041414.1 | - | 7 - 8 | 871 | 134 | |
DA176757.1 | - | 2 - 3 | 183 | 134 |