The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000351.5 | NP_000342.2 | 1 - 2 | 250 | 67 |
GenBank mRNA | AK314034.1 | - | 1 - 2 | 190 | 67 |
J04964.1 | - | 1 - 2 | 208 | 67 | |
M16505.1 | - | 1 - 2 | 225 | 67 |