The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000351.5 | NP_000342.2 | 4 - 5 | 645 | 105 |
NM_001320750.1 | NP_001307679.1 | 5 - 6 | 869 | 105 | |
NM_001320751.1 | NP_001307680.1 | 6 - 7 | 863 | 105 | |
NM_001320752.1 | NP_001307681.1 | 5 - 6 | 709 | 105 | |
NM_001320753.1 | NP_001307682.1 | 4 - 5 | 487 | 105 | |
NM_001320754.1 | NP_001307683.1 | 4 - 5 | 635 | 105 | |
GenBank mRNA | AK314034.1 | - | 4 - 5 | 585 | 105 |
AL549736.3 | - | 4 - 5 | 645 | 105 | |
BC075030.2 | - | 4 - 5 | 421 | 105 | |
J04964.1 | - | 4 - 5 | 603 | 105 | |
M16505.1 | - | 4 - 5 | 620 | 105 |