The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000351.5 | NP_000342.2 | 8 - 9 | 1502 | 96 |
NM_001320750.1 | NP_001307679.1 | 9 - 10 | 1726 | 96 | |
NM_001320751.1 | NP_001307680.1 | 10 - 11 | 1720 | 96 | |
NM_001320752.1 | NP_001307681.1 | 9 - 10 | 1566 | 96 | |
NM_001320753.1 | NP_001307682.1 | 8 - 9 | 1344 | 96 | |
NM_001320754.1 | NP_001307683.1 | 8 - 9 | 1492 | 96 | |
GenBank mRNA | AK314034.1 | - | 8 - 9 | 1442 | 96 |
BC075030.2 | - | 8 - 9 | 1278 | 96 | |
J04964.1 | - | 8 - 9 | 1460 | 96 | |
M16505.1 | - | 8 - 9 | 1477 | 96 | |
M17591.1 | - | 2 - 3 | 192 | 96 |